Product Details

Assay Reference Genome
Location

Chr.19:18786470 on build GRCh38
Cytoband
19p13.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1311
Gene Symbol
COMP
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
Location
Chr.19:18782773-18791305 on build GRCh38
Assay Gene Location
Within Intron 11
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
COMP NM_000095.2 NP_000086.2
AB086984.1 BAC53888.1
AK074508.1 BAC11031.1
AK223216.1 BAD96936.1
AK290595.1
AK296586.1
AK297798.1
AK297814.1
BC110847.1 AAI10848.1
BC125092.1
L32137.1 AAA57253.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3643845 Chr19:18786208 - 18791888 on Build GRCh38 Loss COMP
nsv1151217 Chr19:18627369 - 19284914 on Build GRCh38 Deletion SUGP2 LOC102724360 RFXANK UPF1 BORCS8 KLHL26 CERS1 DDX49 NCAN HAPLN4 SUGP1 GDF1 NR2C2AP CRTC1 BORCS8-MEF2B TMEM161A SLC25A42 COPE MEF2B ARMC6 TM6SF2 HOMER3 COMP
nsv952422 Chr19:18606691 - 18807891 on Build GRCh38 Deletion CRTC1 KLHL26 TMEM59L CRLF1 COMP
nsv833777 Chr19:18765126 - 18920267 on Build GRCh38 Loss GDF1 UPF1 CRTC1 CERS1 COPE DDX49 COMP

View Full Product Details