Product Details

Assay Reference Genome
Location

Chr.20:56422210 on build GRCh38
Cytoband
20q13.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
57091
Gene Symbol
CASS4
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
C20orf32, CAS4, HEFL, HEPL
Location
Chr.20:56411550-56460240 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CASS4 NM_001164114.1 NP_001157586.1
NM_001164115.1 NP_001157587.1
NM_001164116.1 NP_001157588.1
NM_020356.3 NP_065089.2
XM_006723831.3 XP_006723894.1
AJ276678.1 CAC00655.1
AK027760.1 BAB55351.1
AK295613.1
AK308057.1
AK314653.1
BC027951.1 AAH27951.1
BX280195.1
DA584687.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv525425 Chr20:56403979 - 56897294 on Build GRCh38 Gain FAM209B TFAP2C FAM209A CSTF1 LOC105372682 GCNT7 CASS4 RTFDC1
nsv834013 Chr20:56307279 - 56476126 on Build GRCh38 Gain FAM210B AURKA CSTF1 CASS4 RTFDC1
nsv518442 Chr20:56414270 - 56429946 on Build GRCh38 Gain CASS4

View Full Product Details