The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000572.2 | NP_000563.1 | 1 - 2 | 224 | 86 |
GenBank mRNA | AF043333.1 | - | 1 - 2 | 111 | 86 |
AY029171.1 | - | 1 - 2 | 165 | 86 | |
BC104252.1 | - | 1 - 2 | 221 | 86 | |
BC104253.1 | - | 1 - 2 | 221 | 86 | |
CR541993.1 | - | 1 - 2 | 165 | 86 | |
CR542028.1 | - | 1 - 2 | 165 | 86 | |
M57627.1 | - | 1 - 2 | 195 | 86 |