The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243211.1 | NP_001230140.1 | 3 - 4 | 443 | 108 |
NM_001562.3 | NP_001553.1 | 4 - 5 | 455 | 108 | |
XM_011542805.1 | XP_011541107.1 | 3 - 4 | 451 | 108 | |
XM_011542806.2 | XP_011541108.1 | 4 - 5 | 459 | 108 | |
GenBank mRNA | AF380360.1 | - | 4 - 5 | 287 | 108 |
AY044641.1 | - | 4 - 5 | 332 | 108 | |
AY266351.1 | - | 3 - 4 | 400 | 108 | |
BC007007.1 | - | 4 - 5 | 430 | 108 | |
BC007461.1 | - | 4 - 5 | 455 | 108 | |
CR541973.1 | - | 3 - 4 | 235 | 108 | |
CR542001.1 | - | 3 - 4 | 235 | 108 | |
D49950.1 | - | 4 - 5 | 412 | 108 | |
U90434.1 | - | 3 - 4 | 232 | 108 |