The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000389.4 | NP_000380.1 | 2 - 3 | 565 | 99 |
NM_001220777.1 | NP_001207706.1 | 2 - 3 | 510 | 99 | |
NM_001220778.1 | NP_001207707.1 | 2 - 3 | 675 | 99 | |
NM_001291549.1 | NP_001278478.1 | 3 - 4 | 717 | 99 | |
NM_078467.2 | NP_510867.1 | 3 - 4 | 512 | 99 | |
GenBank mRNA | AB209881.1 | - | 2 - 3 | 522 | 99 |
AK298901.1 | - | 2 - 3 | 574 | 99 | |
AK309512.1 | - | 3 - 4 | 411 | 99 | |
BC000275.1 | - | 2 - 3 | 505 | 99 | |
BC000312.2 | - | 2 - 3 | 659 | 99 | |
BC001935.1 | - | 2 - 3 | 520 | 99 | |
BC013967.2 | - | 2 - 3 | 523 | 99 | |
BU154329.1 | - | 3 - 4 | 533 | 99 | |
BU193165.1 | - | 3 - 4 | 717 | 99 | |
DA838987.1 | - | 2 - 3 | 473 | 99 | |
L25610.1 | - | 2 - 3 | 518 | 99 | |
L26165.1 | - | 2 - 3 | 518 | 99 | |
L47232.1 | - | 2 - 3 | 493 | 99 | |
L47233.1 | - | 2 - 3 | 493 | 99 | |
S67388.1 | - | 2 - 3 | 515 | 99 | |
U03106.1 | - | 2 - 3 | 515 | 99 | |
U09579.1 | - | 2 - 3 | 534 | 99 |