The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012092.3 | NP_036224.1 | 4 - 5 | 658 | 90 |
GenBank mRNA | AB023135.1 | - | 4 - 5 | 616 | 90 |
AJ277832.1 | - | 4 - 5 | 658 | 90 | |
BC028210.1 | - | 4 - 5 | 626 | 90 |