The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003005.3 | NP_002996.2 | 8 - 9 | 1402 | 61 |
XM_005245435.1 | XP_005245492.1 | 8 - 9 | 1399 | 61 | |
XM_005245436.3 | XP_005245493.1 | 8 - 9 | 1399 | 61 | |
XM_005245438.1 | XP_005245495.1 | 8 - 9 | 1399 | 61 | |
XM_005245439.1 | XP_005245496.1 | 8 - 9 | 1399 | 61 | |
XM_005245440.1 | XP_005245497.1 | 7 - 8 | 1213 | 61 | |
GenBank mRNA | AK312884.1 | - | 8 - 9 | 1384 | 61 |
BC028067.2 | - | 7 - 8 | 1334 | 61 | |
BC068533.1 | - | 8 - 9 | 1388 | 61 | |
KU178327.1 | - | 7 - 8 | 1337 | 61 | |
M25322.1 | - | 8 - 9 | 1375 | 61 |