The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000181.3 | NP_000172.2 | 11 - 12 | 1925 | 81 |
NM_001284290.1 | NP_001271219.1 | 9 - 10 | 1487 | 81 | |
NM_001293104.1 | NP_001280033.1 | 10 - 11 | 1740 | 81 | |
NM_001293105.1 | NP_001280034.1 | 9 - 10 | 1597 | 81 | |
NR_120531.1 | 10 - 11 | 1840 | 81 | ||
XM_005250297.3 | XP_005250354.1 | 10 - 11 | 1767 | 81 | |
XM_011516113.1 | XP_011514415.1 | 10 - 11 | 1669 | 81 | |
XM_011516114.1 | XP_011514416.1 | 10 - 11 | 1597 | 81 | |
XM_017012091.1 | XP_016867580.1 | 9 - 10 | 1516 | 81 | |
XM_017012092.1 | XP_016867581.1 | 9 - 10 | 1503 | 81 | |
XM_017012093.1 | XP_016867582.1 | 9 - 10 | 1444 | 81 | |
GenBank mRNA | AK096764.1 | - | 3 - 4 | 2183 | 81 |
AK223406.1 | - | 11 - 12 | 1824 | 81 | |
AK300223.1 | - | 11 - 12 | 1714 | 81 | |
AK303819.1 | - | 9 - 10 | 1384 | 81 | |
AK312752.1 | - | 11 - 12 | 1841 | 81 | |
BC014142.2 | - | 11 - 12 | 1795 | 81 | |
BM719712.1 | - | 2 - 3 | 178 | 81 | |
CD014092.1 | - | 10 - 11 | 1622 | 81 | |
CD014093.1 | - | 9 - 10 | 1480 | 81 | |
CD014094.1 | - | 10 - 11 | 1722 | 81 | |
M15182.1 | - | 11 - 12 | 1820 | 81 |