Gene Transcripts
- Gene Symbol
- Pafah2
- Entrez Gene ID
- 100163
- Gene Name
- platelet-activating factor acetylhydrolase 2
- Gene Aliases
- 2310074E22Rik, AI507170
- Chromosome Location
- Chr. 4: 134396320 - 134427412 on Build GRCm38
- UniGene ID
- Mm.22116
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285872.1 |
NP_001272801.1
|
8, 9 |
909 |
|
NM_001285874.1 |
NP_001272803.1
|
7, 8 |
851 |
|
NM_001285875.1 |
NP_001272804.1
|
7, 8 |
801 |
|
NM_001285877.1 |
NP_001272806.1
|
5, 6 |
545 |
|
NM_133880.3 |
NP_598641.2
|
8, 9 |
898 |
|
GenBank mRNA |
AK144090.1 |
- |
8, 9 |
861 |
|
AK145092.1 |
- |
7, 8 |
752 |
|
AK156774.1 |
- |
7, 8 |
852 |
|
AK181929.1 |
- |
1, 2 |
67 |
|
BC021890.1 |
- |
7, 8 |
803 |
3598000 |
BC025495.1 |
- |
8, 9 |
920 |
5310200 |
BC025938.1 |
- |
7, 8 |
744 |
|
BQ179290.1 |
- |
6, 7 |
620 |
|
View Full Product Details