Gene Transcripts

Gene Symbol
Snx25
Entrez Gene ID
102141
Gene Name
sorting nexin 25
Gene Aliases
AI661919, Gm1699, Gm173, Sbbi31
Chromosome Location
Chr. 8: 46033261 - 46151858 on Build GRCm38
UniGene ID
Mm.267258
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207213.2 NP_997096.2 2 232
GenBank mRNA AY601646.1 - 1 451
BF466223.1 - 3 352

View Full Product Details