Gene Transcripts

Gene Symbol
Exoc5
Entrez Gene ID
105504
Gene Name
exocyst complex component 5
Gene Aliases
AI447711, AI448003, Gm76, PRO1912, SEC10, Sec10l1
Chromosome Location
Chr. 14: 49011922 - 49066667 on Build GRCm38
UniGene ID
Mm.31607
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207214.3 NP_997097.1 7 1166
GenBank mRNA AK134562.1 - 7 678
AK152307.1 - 7 895
AK157473.1 - 7 861
AK164067.1 - 7 888
AK167145.1 - 7 854
AK170148.1 - 7 859
AK171012.1 - 7 884
BC049967.1 - 7 823 3672313

View Full Product Details