Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033226.4 | NP_001028398.1 | 7 | 1070 | |
NM_001285895.1 | NP_001272824.1 | 8 | 1034 | ||
NM_001285896.1 | NP_001272825.1 | 8 | 1047 | ||
NM_001285897.1 | NP_001272826.1 | 8 | 1289 | ||
NM_001285898.1 | NP_001272827.1 | 8 | 1299 | ||
GenBank mRNA | AK077094.1 | - | 8 | 1139 | |
AK139540.1 | - | 8 | 1049 | ||
AK163350.1 | - | 7 | 1071 | ||
BC044740.2 | - | 8 | 1299 | ||
JN255799.1 | - | 8 | 1034 |