Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289730.1 | NP_001276659.1 | 3 | 371 | |
NM_001289731.1 | NP_001276660.1 | 6 | 927 | ||
NM_133197.4 | NP_573460.2 | 3 | 371 | ||
GenBank mRNA | AB052945.1 | - | 3 | 314 | |
AB101616.1 | - | 3 | 314 | ||
AK032122.1 | - | 3 | 372 | ||
AK045424.1 | - | 6 | 929 | ||
BC125435.1 | - | 3 | 273 | 40129950 |