Gene Transcripts

Gene Symbol
Vmn1r11
Entrez Gene ID
113860
Gene Name
vomeronasal 1 receptor 11
Gene Aliases
V1rc3
Chromosome Location
Chr. 6: 57137353 - 57138252 on Build GRCm38
UniGene ID
Mm.377157
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053233.2 NP_444463.2 1 488
GenBank mRNA BC104398.1 - 1 488
BC104399.1 - 1 488 40050460

View Full Product Details