Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053268.2 | NP_444498.2 | 10 | 925 | |
GenBank mRNA | AB056433.1 | - | 10 | 918 | |
AK034434.1 | - | 10 | 925 | ||
AK082530.1 | - | 1 | 77 | ||
AK154499.1 | - | 10 | 942 | ||
AK166654.1 | - | 10 | 925 |