Gene Transcripts

Gene Symbol
Acvr2b
Entrez Gene ID
11481
Gene Name
activin receptor IIB
Gene Aliases
ActRIIB
Chromosome Location
Chr. 9: 119401583 - 119442134 on Build GRCm38
UniGene ID
Mm.390239
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007397.2 NP_031423.1 10 1560
GenBank mRNA AK137432.1 - 10 1312
BC106189.1 - 11 1545 30240596
M84120.1 - 10 1558

View Full Product Details