Gene Transcripts

Gene Symbol
Agxt
Entrez Gene ID
11611
Gene Name
alanine-glyoxylate aminotransferase
Gene Aliases
AGT, Agt1, Agxt1, SPT
Chromosome Location
Chr. 1: 93135240 - 93145421 on Build GRCm38
UniGene ID
Mm.7457
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276710.1 NP_001263639.1 2 421
NM_016702.3 NP_057911.2 2 421
GenBank mRNA AF027730.2 - 2 416
AK149432.1 - 2 344
BC025799.1 - 2 385 5099627

View Full Product Details