Gene Transcripts

Gene Symbol
Lsm10
Entrez Gene ID
116748
Gene Name
U7 snRNP-specific Sm-like protein LSM10
Gene Aliases
Chromosome Location
Chr. 4: 126096577 - 126098584 on Build GRCm38
UniGene ID
Mm.41865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163266.1 NP_001156738.1 2 342
NM_138721.2 NP_620046.1 2 444
GenBank mRNA AK143443.1 - 2 383
BC024543.1 - 2 345 4989202
BC026476.1 - 2 373 4489522
BE944742.1 - 1 480

View Full Product Details