Gene Transcripts
- Gene Symbol
- Bckdk
- Entrez Gene ID
- 12041
- Gene Name
- branched chain ketoacid dehydrogenase kinase
- Gene Aliases
- AI327402
- Chromosome Location
- Chr. 7: 127904073 - 127909664 on Build GRCm38
- UniGene ID
- Mm.425495
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009739.3 |
NP_033869.1
|
11, 12 |
1354 |
|
GenBank mRNA |
AF043070.1 |
- |
10, 11 |
1195 |
|
AK075602.1 |
- |
11, 12 |
1280 |
|
AK145479.1 |
- |
11, 12 |
1325 |
|
AK150607.1 |
- |
11, 12 |
1346 |
|
AK150723.1 |
- |
11, 12 |
1346 |
|
AK167519.1 |
- |
11, 12 |
1346 |
|
AK207870.1 |
- |
2, 3 |
199 |
|
BC004077.1 |
- |
8, 9 |
782 |
|
BC046595.1 |
- |
11, 12 |
1324 |
6313106 |
View Full Product Details