Gene Transcripts

Gene Symbol
Bgn
Entrez Gene ID
12111
Gene Name
biglycan
Gene Aliases
BG, DSPG1, PG-S1, PGI, SLRR1A
Chromosome Location
Chr. X: 73483612 - 73495936 on Build GRCm38
UniGene ID
Mm.2608
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007542.4 NP_031568.2 2 196
GenBank mRNA AK086920.1 - 2 193
AK135496.1 - 2 197
AK155973.1 - 2 197
AK164867.1 - 2 197
AK171870.1 - 2 197
BC005452.1 - 2 190 3594469
BC019502.1 - 2 199 4237268
BC052857.1 - 2 181 30061850
BC057185.1 - 2 208
L20276.1 - 2 196
X53928.1 - 291
Y11758.1 - 1 27

View Full Product Details