Gene Transcripts

Gene Symbol
Abcc2
Entrez Gene ID
12780
Gene Name
ATP-binding cassette, sub-family C (CFTR/MRP), member 2
Gene Aliases
AI173996, Abc30, Cmoat, Mrp2, cMRP
Chromosome Location
Chr. 19: 43782308 - 43838332 on Build GRCm38
UniGene ID
Mm.39054
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013806.2 NP_038834.2 23 3268
GenBank mRNA AF227274.1 - 23 3172
AF282772.1 - 23 3172
AF282773.1 - 23 3172

View Full Product Details