Gene Transcripts

Gene Symbol
Col11a2
Entrez Gene ID
12815
Gene Name
collagen, type XI, alpha 2
Gene Aliases
Chromosome Location
Chr. 17: 34038486 - 34066684 on Build GRCm38
UniGene ID
Mm.20230
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009926.1 NP_034056.1 61 4591
GenBank mRNA KJ650220.1 - 62 4568
U16789.1 - 61 4592

View Full Product Details