Gene Transcripts
- Gene Symbol
- Csn1s2b
- Entrez Gene ID
- 12992
- Gene Name
- casein alpha s2-like B
- Gene Aliases
- AW987150, Csnd, Csne
- Chromosome Location
- Chr. 5: 87808121 - 87824421 on Build GRCm38
- UniGene ID
- Mm.153308
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301334.1 |
NP_001288263.1
|
2, 3 |
99 |
|
NM_001301335.1 |
NP_001288264.1
|
2, 3 |
99 |
|
NM_009973.3 |
NP_034103.2
|
2, 3 |
99 |
|
GenBank mRNA |
AK021337.1 |
- |
2, 3 |
95 |
|
AK021343.1 |
- |
2, 3 |
97 |
|
AK052780.1 |
- |
2, 3 |
97 |
|
AK085744.1 |
- |
2, 3 |
99 |
|
AK085747.1 |
- |
2, 3 |
97 |
|
AK164763.1 |
- |
2, 3 |
97 |
|
AK164767.1 |
- |
2, 3 |
99 |
|
AK164768.1 |
- |
2, 3 |
97 |
|
AK164795.1 |
- |
2, 3 |
95 |
|
BC002084.1 |
- |
2, 3 |
80 |
3489886 |
J00379.1 |
- |
1, 2 |
65 |
|
V00740.1 |
- |
1, 2 |
65 |
|
View Full Product Details