Gene Transcripts

Gene Symbol
Cyp2j6
Entrez Gene ID
13110
Gene Name
cytochrome P450, family 2, subfamily j, polypeptide 6
Gene Aliases
Chromosome Location
Chr. 4: 96516138 - 96560835 on Build GRCm38
UniGene ID
Mm.98200
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010008.4 NP_034138.3 4, 5 876
GenBank mRNA AK033614.1 - 4, 5 879
AK045421.1 - 4, 5 866
AK084025.1 - 4, 5 886
AK165663.1 - 4, 5 721
BC050832.1 - 4, 5 870 6514145
U62295.1 - 4, 5 923

View Full Product Details