Gene Transcripts
- Gene Symbol
- Ebf3
- Entrez Gene ID
- 13593
- Gene Name
- early B cell factor 3
- Gene Aliases
- 3110018A08Rik, O/E-2, mKIAA4201
- Chromosome Location
- Chr. 7: 137193671 - 137322810 on Build GRCm38
- UniGene ID
- Mm.258708
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113414.1 |
NP_001106885.1
|
7, 8 |
1169 |
|
NM_001113415.1 |
NP_001106886.1
|
7, 8 |
1169 |
|
NM_010096.3 |
NP_034226.1
|
7, 8 |
1169 |
|
GenBank mRNA |
AK138602.1 |
- |
2, 3 |
358 |
|
AK220542.1 |
- |
7, 8 |
1169 |
|
BC067018.1 |
- |
8, 9 |
924 |
6833120 |
CF727275.1 |
- |
1, 2 |
34 |
|
U82442.1 |
- |
6, 7 |
466 |
|
U92702.1 |
- |
7, 8 |
710 |
|
U92704.1 |
- |
7, 8 |
710 |
|
View Full Product Details