Gene Transcripts

Gene Symbol
Eln
Entrez Gene ID
13717
Gene Name
elastin
Gene Aliases
AI385707, AI480567, E030024M20Rik
Chromosome Location
Chr. 5: 134702596 - 134747367 on Build GRCm38
UniGene ID
Mm.275320
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007925.3 NP_031951.2 7 380
GenBank mRNA AK041860.1 - 7 381
AK080277.1 - 7 386
BC051074.1 - 7 372
BC051649.1 - 7 449 5346255

View Full Product Details