Gene Transcripts

Gene Symbol
Ephb6
Entrez Gene ID
13848
Gene Name
Eph receptor B6
Gene Aliases
Cekl, Mep
Chromosome Location
Chr. 6: 41605040 - 41620507 on Build GRCm38
UniGene ID
Mm.271976
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146351.1 NP_001139823.1 15 2992
NM_007680.4 NP_031706.3 15 2988
GenBank mRNA AK076442.1 - 15 2815
AK087423.1 - 15 2903
AK163823.1 - 6 710
BC031924.1 - 14 2600 4911959
L77867.1 - 15 2682

View Full Product Details