Gene Transcripts

Gene Symbol
Ext1
Entrez Gene ID
14042
Gene Name
exostoses (multiple) 1
Gene Aliases
AA409028
Chromosome Location
Chr. 15: 53068261 - 53346183 on Build GRCm38
UniGene ID
Mm.309395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010162.2 NP_034292.2 8 2528
GenBank mRNA AK132327.1 - 8 2505
BC004741.1 - 8 1892 3499743
U78539.1 - 8 2032
X96639.1 - 8 2299

View Full Product Details