Gene Transcripts
- Gene Symbol
- Ganab
- Entrez Gene ID
- 14376
- Gene Name
- alpha glucosidase 2 alpha neutral subunit
- Gene Aliases
- AU042638, G2an, GluII, mKIAA0088
- Chromosome Location
- Chr. 19: 8898057 - 8916742 on Build GRCm38
- UniGene ID
- Mm.3196
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001293621.1 |
NP_001280550.1
|
18, 19 |
2312 |
|
NM_008060.2 |
NP_032086.1
|
19, 20 |
2378 |
|
GenBank mRNA |
AK017873.1 |
- |
6, 7 |
718 |
|
AK028655.1 |
- |
18, 19 |
2282 |
|
AK030722.1 |
- |
18, 19 |
2281 |
|
AK081915.1 |
- |
18, 19 |
2274 |
|
AK122201.1 |
- |
13, 14 |
2508 |
|
AK166150.1 |
- |
18, 19 |
2251 |
|
BC020386.1 |
- |
11, 12 |
1528 |
|
BC035423.1 |
- |
9, 10 |
1128 |
|
BC094437.1 |
- |
18, 19 |
2262 |
30682580 |
BC117888.1 |
- |
18, 19 |
2255 |
40096413 |
BC117889.1 |
- |
18, 19 |
2258 |
40096414 |
BC128069.1 |
- |
19, 20 |
2322 |
40096428 |
U92793.1 |
- |
19, 20 |
2324 |
|
View Full Product Details