Gene Transcripts

Gene Symbol
G6pc
Entrez Gene ID
14377
Gene Name
glucose-6-phosphatase, catalytic
Gene Aliases
AW107337, G6Pase, G6pt, Glc-6-Pase
Chromosome Location
Chr. 11: 101367730 - 101377903 on Build GRCm38
UniGene ID
Mm.18064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008061.3 NP_032087.2 4 584
GenBank mRNA AK050279.1 - 4 599
AK052656.1 - 4 599
BC013448.1 - 4 584 4237115
U00445.1 - 4 599

View Full Product Details