Gene Transcripts

Gene Symbol
Opn1mw
Entrez Gene ID
14539
Gene Name
opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan)
Gene Aliases
Gcp
Chromosome Location
Chr. X: 74127466 - 74150756 on Build GRCm38
UniGene ID
Mm.284825
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008106.2 NP_032132.1 1 84
GenBank mRNA AF011389.1 - 1 70
AF190672.1 - 1 86
AK149287.1 - 1 85
BC014826.1 - 1 64 4507311

View Full Product Details