Gene Transcripts

Gene Symbol
Gdf11
Entrez Gene ID
14561
Gene Name
growth differentiation factor 11
Gene Aliases
BMP-11, Bmp11
Chromosome Location
Chr. 10: 128884546 - 128891718 on Build GRCm38
UniGene ID
Mm.299218
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010272.1 NP_034402.1 2 615
GenBank mRNA AF092734.1 - 2 360
KC561772.1 - 1727

View Full Product Details