Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001160406.1 | NP_001153878.1 | 4 | 629 | |
NM_008114.3 | NP_032140.1 | 4 | 629 | ||
GenBank mRNA | AF017275.1 | - | 4 | 588 | |
AK157262.1 | - | 4 | 638 | ||
BC052654.1 | - | 4 | 604 | 30039774 | |
BC132365.1 | - | 4 | 565 | 40130642 | |
BC132367.1 | - | 4 | 565 | 40130644 | |
BC145238.1 | - | 4 | 565 | 9053790 |