Gene Transcripts

Gene Symbol
Gna12
Entrez Gene ID
14673
Gene Name
guanine nucleotide binding protein, alpha 12
Gene Aliases
AI414047, AI504261, Galpha12
Chromosome Location
Chr. 5: 140759944 - 140830431 on Build GRCm38
UniGene ID
Mm.370185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010302.2 NP_034432.1 4 975
GenBank mRNA AK194048.1 - 3 373
BC111810.1 - 5 742 40045663
M57618.1 - 1 150
M63659.1 - 4 975

View Full Product Details