Gene Transcripts

Gene Symbol
Gp5
Entrez Gene ID
14729
Gene Name
glycoprotein 5 (platelet)
Gene Aliases
GPV
Chromosome Location
Chr. 16: 30307685 - 30310781 on Build GRCm38
UniGene ID
Mm.3519
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008148.4 NP_032174.2 2 311
GenBank mRNA AK031272.1 - 2 310
AK172059.1 - 2 310

View Full Product Details