Gene Transcripts

Gene Symbol
Gpr33
Entrez Gene ID
14762
Gene Name
G protein-coupled receptor 33
Gene Aliases
Chromosome Location
Chr. 12: 52023003 - 52028063 on Build GRCm38
UniGene ID
Mm.12890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008159.2 NP_032185.1 2 149
GenBank mRNA AF045766.1 - 2 172
BC064084.1 - 2 149 575313
GQ981323.1 - 2 190
GQ981327.1 - 2 252

View Full Product Details