Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008214.4 | NP_032240.3 | 11 | 1384 | |
GenBank mRNA | AK011685.1 | - | 11 | 1300 | |
AK088824.1 | - | 11 | 1337 | ||
AK146922.1 | - | 11 | 1337 | ||
AK150081.1 | - | 11 | 1303 | ||
AK152329.1 | - | 11 | 1316 | ||
AK166826.1 | - | 11 | 1300 | ||
AK166892.1 | - | 11 | 1347 | ||
BC020088.1 | - | 11 | 1319 | 4008479 | |
U39473.1 | - | 11 | 1364 |