Gene Transcripts
- Gene Symbol
- Inppl1
- Entrez Gene ID
- 16332
- Gene Name
- inositol polyphosphate phosphatase-like 1
- Gene Aliases
- 51C, SHIP2
- Chromosome Location
- Chr. 7: 101822632 - 101838226 on Build GRCm38
- UniGene ID
- Mm.476000
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001122739.1 |
NP_001116211.1
|
24, 25 |
2959 |
|
NM_010567.2 |
NP_034697.2
|
24, 25 |
3193 |
|
GenBank mRNA |
AF162781.1 |
- |
24, 25 |
3043 |
|
AK133359.1 |
- |
24, 25 |
2920 |
|
AK134307.1 |
- |
9, 10 |
915 |
|
AK149873.1 |
- |
24, 25 |
2977 |
|
AK187812.1 |
- |
1, 2 |
37 |
|
BC039207.1 |
- |
11, 12 |
1095 |
|
BC063080.1 |
- |
24, 25 |
2889 |
6416946 |
U92477.1 |
- |
5, 6 |
545 |
|
View Full Product Details