Gene Transcripts
- Gene Symbol
- Lamc2
- Entrez Gene ID
- 16782
- Gene Name
- laminin, gamma 2
- Gene Aliases
- AA589349
- Chromosome Location
- Chr. 1: 153122756 - 153186447 on Build GRCm38
- UniGene ID
- Mm.329272
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008485.3 |
NP_032511.3
|
15 |
2377 |
|
GenBank mRNA |
U43327.2 |
- |
14 |
2107 |
|
View Full Product Details