Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017959.2 | NP_001017959.1 | 4 | 635 | |
NM_001290485.1 | NP_001277414.1 | 4 | 635 | ||
NM_010685.4 | NP_034815.2 | 4 | 635 | ||
GenBank mRNA | AK012149.1 | - | 4 | 594 | |
AK032974.1 | - | 4 | 597 | ||
AK048174.1 | - | 4 | 577 | ||
AK051989.1 | - | 3 | 1433 | ||
AK078193.1 | - | 4 | 629 | ||
AK088804.1 | - | 5 | 612 | ||
AK128946.1 | - | 2 | 297 | ||
AK136550.1 | - | 4 | 595 | ||
AK146731.1 | - | 4 | 616 | ||
AK150766.1 | - | 4 | 604 | ||
AK159272.1 | - | 4 | 590 | ||
AK159731.1 | - | 4 | 590 | ||
AK163933.1 | - | 4 | 636 | ||
AK217258.1 | - | 2 | 243 | ||
BC138718.1 | - | 4 | 522 | 8861740 | |
BC138723.1 | - | 4 | 522 | 8861745 | |
J05287.1 | - | 4 | 562 |