Gene Transcripts

Gene Symbol
Vmn1r66
Entrez Gene ID
171264
Gene Name
vomeronasal 1 receptor 66
Gene Aliases
S67, V1re11, V1re3
Chromosome Location
Chr. 7: 10273828 - 10275351 on Build GRCm38
UniGene ID
Mm.222637
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134230.3 NP_598991.2 1 766
GenBank mRNA AF394952.1 - 1 765
BC109341.1 - 1 619 40052743
BC109342.1 - 1 619 40052744

View Full Product Details