Gene Transcripts
- Gene Symbol
- Ccndbp1
- Entrez Gene ID
- 17151
- Gene Name
- cyclin D-type binding-protein 1
- Gene Aliases
- AU022347, DIP1, GCIP, Maid, SECC-8, SSEC-8
- Chromosome Location
- Chr. 2: 121008383 - 121016912 on Build GRCm38
- UniGene ID
- Mm.7838
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010761.2 |
NP_034891.2
|
2, 3 |
270 |
|
GenBank mRNA |
AK144883.1 |
- |
2, 3 |
247 |
|
AK154285.1 |
- |
2, 3 |
224 |
|
AK160205.1 |
- |
2, 3 |
236 |
|
AK163150.1 |
- |
2, 3 |
183 |
|
AK167345.1 |
- |
2, 3 |
235 |
|
AK186314.1 |
- |
1, 2 |
47 |
|
BC001984.1 |
- |
2, 3 |
206 |
3489973 |
BC005802.1 |
- |
1, 2 |
279 |
3707919 |
CO810054.1 |
- |
2, 3 |
277 |
|
U50734.1 |
- |
2, 3 |
188 |
|
View Full Product Details