Gene Transcripts
- Gene Symbol
- Mre11a
- Entrez Gene ID
- 17535
- Gene Name
- meiotic recombination 11 homolog A (S. cerevisiae)
- Gene Aliases
- Mre11, Mre11b
- Chromosome Location
- Chr. 9: 14784670 - 14837125 on Build GRCm38
- UniGene ID
- Mm.149071
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018736.2 |
NP_061206.1
|
2, 3 |
155 |
|
GenBank mRNA |
AK041248.1 |
- |
2, 3 |
154 |
|
AK141214.1 |
- |
2, 3 |
167 |
|
AK146234.1 |
- |
2, 3 |
188 |
|
AK146370.1 |
- |
2, 3 |
188 |
|
AK168180.1 |
- |
2, 3 |
188 |
|
AK168208.1 |
- |
2, 3 |
189 |
|
BC065144.1 |
- |
2, 3 |
155 |
30627207 |
U58987.1 |
- |
2, 3 |
190 |
|
U60318.1 |
- |
2, 3 |
190 |
|
View Full Product Details