Gene Transcripts

Gene Symbol
Pdha1
Entrez Gene ID
18597
Gene Name
pyruvate dehydrogenase E1 alpha 1
Gene Aliases
Pdha-1
Chromosome Location
Chr. X: 160122219 - 160138336 on Build GRCm38
UniGene ID
Mm.34775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008810.2 NP_032836.1 8 826
GenBank mRNA AK145718.1 - 8 904
AK148459.1 - 8 847
AK167987.1 - 8 852
AK168152.1 - 8 876
AK169289.1 - 8 901
BC007142.1 - 8 826 3600605
M76727.1 - 8 913

View Full Product Details