Gene Transcripts

Gene Symbol
Enpp1
Entrez Gene ID
18605
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 1
Gene Aliases
4833416E15Rik, AI428932, C76301, CD203c, E-NPP 1, E-NPP1, Ly-41, M6S1, NPP1, Npps, PC-1, Pca, Pca-1, Pdnp1, ttw, twy
Chromosome Location
Chr. 10: 24637914 - 24712154 on Build GRCm38
UniGene ID
Mm.27254
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF339910.1 - 24 2511
AK029600.1 - 24 2420
AK041894.1 - 19 1881
AK088857.1 - 24 2515
AK145233.1 - 24 2512
AK164619.1 - 24 2490
BC050011.1 - 24 2662
J02700.2 - 24 2619

View Full Product Details