Gene Transcripts

Gene Symbol
Phkg1
Entrez Gene ID
18682
Gene Name
phosphorylase kinase gamma 1
Gene Aliases
Phkg
Chromosome Location
Chr. 5: 129863435 - 129879083 on Build GRCm38
UniGene ID
Mm.3159
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011079.2 NP_035209.1 9 1052
GenBank mRNA BC055102.1 - 9 1052
J03293.1 - 9 922
M16216.1 - 8 890

View Full Product Details