Gene Transcripts
- Gene Symbol
- Pip5k1c
- Entrez Gene ID
- 18717
- Gene Name
- phosphatidylinositol-4-phosphate 5-kinase, type 1 gamma
- Gene Aliases
- AI115456, AI835305, Pip5kIgamma
- Chromosome Location
- Chr. 10: 81292962 - 81319974 on Build GRCm38
- UniGene ID
- Mm.24222
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001146687.2 |
NP_001140159.1
|
8, 9 |
1234 |
|
NM_001293646.1 |
NP_001280575.1
|
8, 9 |
1234 |
|
NM_001293647.1 |
NP_001280576.1
|
8, 9 |
1247 |
|
NM_008844.3 |
NP_032870.2
|
8, 9 |
1234 |
|
GenBank mRNA |
AB006916.1 |
- |
8, 9 |
1217 |
|
AK014403.1 |
- |
8, 9 |
1200 |
|
AK041402.1 |
- |
8, 9 |
1220 |
|
AK154816.1 |
- |
8, 9 |
1226 |
|
AK155371.1 |
- |
8, 9 |
1210 |
|
AK170286.1 |
- |
8, 9 |
1226 |
|
AK171576.1 |
- |
8, 9 |
1208 |
|
AK182986.1 |
- |
1, 2 |
97 |
|
BC019138.1 |
- |
8, 9 |
1183 |
5041431 |
BC094665.1 |
- |
8, 9 |
1206 |
30356181 |
View Full Product Details