Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172695.2 | NP_766283.2 | 9 | 1508 | |
GenBank mRNA | AK019070.1 | - | 9 | 1514 | |
AK075934.1 | - | 9 | 1440 | ||
AK166122.1 | - | 9 | 1414 | ||
BC139355.1 | - | 9 | 1507 | 8862377 | |
BC139356.1 | - | 9 | 1507 | 8862378 | |
BC139773.1 | - | 9 | 1418 | 40111789 |