Gene Transcripts
- Gene Symbol
- Prrx1
- Entrez Gene ID
- 18933
- Gene Name
- paired related homeobox 1
- Gene Aliases
- A230024N07Rik, AA755424, AI385634, AI843499, K-2, Pmx, Pmx1, Prx1, mHox
- Chromosome Location
- Chr. 1: 163245119 - 163313705 on Build GRCm38
- UniGene ID
- Mm.288642
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001025570.1 |
NP_001020741.1
|
1, 2 |
1254 |
|
NM_011127.2 |
NP_035257.1
|
1, 2 |
1254 |
|
NM_175686.3 |
NP_783617.2
|
1, 2 |
1254 |
|
GenBank mRNA |
AB221686.1 |
- |
1, 2 |
239 |
|
AK019503.1 |
- |
1, 2 |
65 |
|
AK031075.1 |
- |
1, 2 |
220 |
|
AK032773.1 |
- |
1, 2 |
821 |
|
AK084387.1 |
- |
1, 2 |
1272 |
|
AK089006.1 |
- |
1, 2 |
1086 |
|
AK165914.1 |
- |
1, 2 |
27 |
|
BC092372.1 |
- |
1, 2 |
603 |
30533016 |
L06502.1 |
- |
1, 2 |
419 |
|
U03873.1 |
- |
1, 2 |
1260 |
|
X59725.1 |
- |
1, 2 |
380 |
|
View Full Product Details