Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011132.2 | NP_035262.2 | 25 | 3110 | |
GenBank mRNA | AF123502.1 | - | 25 | 3084 | |
AK141674.1 | - | 25 | 3071 | ||
BC048166.1 | - | 25 | 3045 | 6410453 | |
BC063246.1 | - | 25 | 3045 | 6410453 |